Learn More
Description
This peptide corresponds to 15 amino acids near the middle of WNV Envelope Protein.
Specifications
Specifications
| Species | Virus |
| Purification Method | purified |
| Common Name | West Nile Virus Envelope |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| For Use With (Application) | Blocking Assay, Control |
| Gene Alias | West Nile envelope; West Nile Virus; WNV envelope |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.