Learn More
World Precision Instrument Westcott Curved Scissors, 11cm, Curved, Blunt, Titanium
SDP
Catalog No. 502538317
Encompass
Titanium Westcott Spring Scissors are popular spring handled scissors that automatically reopen after each cut. Westcott brand spring scissors give you an ergonomic and comfortable grip, reducing hand strain and fatigue during repetitive cutting tasks. Westcott spring scissors are perfect for ambidextrous use, often preferred by left-handed individuals. The tips of these surgical scissors are curved with 19 mm long blades, and they are available with blunt or sharp tips. Our Westcott spring scissors are constructed from titanium, which is 40% lighter than stainless steel, reducing hand fatigue. Titanium scissors are non-magnetic, making them perfect for MRI applications. Titanium is corrosion resistant, an excellent choice for environments with nitric acid, chloride, saltwater, and industrial and organic chemicals.
Catalog No. 50-253-8317 Supplier World Precision Instrument Supplier No. WP3100
Titanium Westcott Spring Scissors are popular spring handled scissors that automatically reopen after each cut. Westcott brand spring scissors give you an ergonomic and comfortable grip, reducing hand strain and fatigue during repetitive cutting tasks. Westcott spring scissors are perfect for ambidextrous use, often preferred by left-handed individuals. The tips of these surgical scissors are curved with 19 mm long blades, and they are available with blunt or sharp tips. Our Westcott spring scissors are constructed from titanium, which is 40% lighter than stainless steel, reducing hand fatigue. Titanium scissors are non-magnetic, making them perfect for MRI applications. Titanium is corrosion resistant, an excellent choice for environments with nitric acid, chloride, saltwater, and industrial and organic chemicals.