Learn More
World Precision Instrument Westcott Student Scissors, 11cm, Blunt Tips
SDP
Catalog No. 502521289
Encompass
Westcott Student Spring Scissors are popular spring handled scissors that automatically reopen after each cut. Westcott brand spring scissors give you an ergonomic and comfortable grip, reducing hand strain and fatigue during repetitive cutting tasks. Westcott spring scissors are perfect for ambidextrous use, often preferred by left-handed individuals. The tips of these surgical scissors are slightly curved, and they are available with blunt or sharp tips. These Westcott scissors are perfect for student use. They are constructed of surgical grade stainless steel and are suitable for autoclaving or chemical sterilization.
Catalog No. 50-252-1289 Supplier World Precision Instrument Supplier No. 503222
Westcott Student Spring Scissors are popular spring handled scissors that automatically reopen after each cut. Westcott brand spring scissors give you an ergonomic and comfortable grip, reducing hand strain and fatigue during repetitive cutting tasks. Westcott spring scissors are perfect for ambidextrous use, often preferred by left-handed individuals. The tips of these surgical scissors are slightly curved, and they are available with blunt or sharp tips. These Westcott scissors are perfect for student use. They are constructed of surgical grade stainless steel and are suitable for autoclaving or chemical sterilization.