Learn More
Bio Rad Laboratories Precision Plus Protein WesternC Blotting Standards, 1.25 ml, molecular weight range 10-250 kD, 10 recombinant proteins, 3 pink reference bands 25 kD, 50 kD, 75 kD
Catalog No. NC1968366
Encompass
Precision Plus Protein WesternC Blotting Standards, 1.25 ml, molecular weight range 10-250 kD, 10 recombinant proteins, 3 pink reference bands 25 kD, 50 kD, 75 kD
Catalog No. NC1968366 Supplier Bio Rad Laboratories Supplier No. 1610399
Precision Plus Protein WesternC Blotting Standards, 1.25 ml, molecular weight range 10-250 kD, 10 recombinant proteins, 3 pink reference bands 25 kD, 50 kD, 75 kD