Learn More
Sigma Aldrich Fine Chemicals Biosciences Western BlockerTM Solution
Catalog No. 502900451
Encompass
Western Blocker SOLUTIONon is used to block both PVDf and nitrocellulose membranes after proteins have been transferred onto them through dot blotting or Western blotting blocking is performed to reduce or eliminate nonspecific binding of primary or secondary antibodies.
Catalog No. 50-290-0451 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W0138400ML
Western Blocker SOLUTIONon is used to block both PVDf and nitrocellulose membranes after proteins have been transferred onto them through dot blotting or Western blotting blocking is performed to reduce or eliminate nonspecific binding of primary or secondary antibodies.