Learn More
Bioworld Western Blot Blocker, 500 g
Catalog No. 50198552
Encompass_Preferred
For those that prefer animal protein as the blocking agent for your immuno-detection analysis. Modified milk protein that is highly soluble and does not inhibit peroxidase detection. The modified milk protein has high blocking efficiency with a clear background.Features:Readily soluble and produces semi-clear solution.No inhibition to peroxidase.Produces clear background.Higher blocking efficiency.Blocking time 30-60 minutes.Fat free.
Catalog No. 50-198-552 Supplier Bioworld Supplier No. 209600072
For those that prefer animal protein as the blocking agent for your immuno-detection analysis. Modified milk protein that is highly soluble and does not inhibit peroxidase detection. The modified milk protein has high blocking efficiency with a clear background.Features:Readily soluble and produces semi-clear solution.No inhibition to peroxidase.Produces clear background.Higher blocking efficiency.Blocking time 30-60 minutes.Fat free.