Learn More
Bioworld Western Blot Box (8.9x6.5x2.5cm)
Catalog No. NC1686204
Encompass_Preferred
bioWORLD's blot boxes are specially designed westerns to fit blots of any size, including whole blots, half blots, and strips. The blot boxes are constructed of ultra-clear, durable, polystyrene plastic with a flat, smooth inner surface and hinged lids to reduce evaporation.
Catalog No. NC1686204 Supplier Bioworld Supplier No. 209300361
bioWORLD's blot boxes are specially designed westerns to fit blots of any size, including whole blots, half blots, and strips. The blot boxes are constructed of ultra-clear, durable, polystyrene plastic with a flat, smooth inner surface and hinged lids to reduce evaporation.