Learn More
LICORbio Black Western Blot Incubation Boxes (large - 1 box)
Catalog No. NC9356877
Encompass_Preferred
Black blot boxes are suitable for incubating near-infrared fluorescent and chemiluminescent Western blots or protein gels. The Large size is: 11.5 cm x 8.8 cm x 2.8 cm.
Catalog No. NC9356877 Supplier LICORbio Supplier No. 92997301
Black blot boxes are suitable for incubating near-infrared fluorescent and chemiluminescent Western blots or protein gels. The Large size is: 11.5 cm x 8.8 cm x 2.8 cm.