Learn More
Electron Microscopy Sciences Western Blotting Boxes Large, Clear, 6 ml, 4⅝x 3 ½ x 1⅛"
SDP
Catalog No. 501929214
Encompass_Preferred
No matter what size your Western blot is, there's a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at a 90 deg angle and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Equipped with an easy-to-open hinged lid trhat reduces evaporation. Applications: Combrex PAGEr Gold, Mighty Small, Novex, SureLock mini.
Color: Clear
Max volume: 6 ml
Dimensions: 4⅝x 3 ½ x 1⅛"
Catalog No. 50-192-9214 Supplier Electron Microscopy Sciences Supplier No. 7047507
No matter what size your Western blot is, there's a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at a 90 deg angle and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Equipped with an easy-to-open hinged lid trhat reduces evaporation. Applications: Combrex PAGEr Gold, Mighty Small, Novex, SureLock mini.
Color: Clear
Max volume: 6 ml
Dimensions: 4⅝x 3 ½ x 1⅛"