Learn More
LICORbio Black Western Blot Incubation Boxes (medium - 5 boxes)
Catalog No. NC9327986
Encompass_Preferred
Black blot boxes are suitable for incubating near-infrared fluorescent and chemiluminescent Western blots or protein gels. The Medium size is: 8.9 cm x 6.5 cm x 2.8 cm.
Catalog No. NC9327986 Supplier LICORbio Supplier No. 92997205
Black blot boxes are suitable for incubating near-infrared fluorescent and chemiluminescent Western blots or protein gels. The Medium size is: 8.9 cm x 6.5 cm x 2.8 cm.