Learn More
Electron Microscopy Sciences Western Blotting Boxes Micro Strip, Clear, 1 ml, 2⅞x 1-3/16 x ¾"
SDP
Catalog No. 501929208
Encompass_Preferred
No matter what size your Western blot is, there's a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at a 90 deg angle and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Equipped with an easy-to-open hinged lid trhat reduces evaporation. Applications: various.
Color: Clear
Max volume: 1 ml
Dimensions: 2⅞x 1-3/16 x ¾"
Catalog No. 50-192-9208 Supplier Electron Microscopy Sciences Supplier No. 7047501
No matter what size your Western blot is, there's a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at a 90 deg angle and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Equipped with an easy-to-open hinged lid trhat reduces evaporation. Applications: various.
Color: Clear
Max volume: 1 ml
Dimensions: 2⅞x 1-3/16 x ¾"