Learn More
Electron Microscopy Sciences Western Blotting Boxes Mini Rectangle, Clear, 3 ml, 2⅞x 2 x 1¼"
SDP
Catalog No. 501929209
Encompass_Preferred
No matter what size your Western blot is, there's a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at a 90 deg angle and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Equipped with an easy-to-open hinged lid trhat reduces evaporation. Applications: half mini protein gels.
Color: Clear
Max volume: 3 ml
Dimensions: 2⅞x 2 x 1¼"
Catalog No. 50-192-9209 Supplier Electron Microscopy Sciences Supplier No. 7047502
No matter what size your Western blot is, there's a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at a 90 deg angle and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Equipped with an easy-to-open hinged lid trhat reduces evaporation. Applications: half mini protein gels.
Color: Clear
Max volume: 3 ml
Dimensions: 2⅞x 2 x 1¼"