Learn More
Sigma Aldrich Fine Chemicals Biosciences Western Blot Box size XXL
Catalog No. 502900543
Encompass
Perfect box for every blotReduce antibody usageHinged Lid minimizes evaporationfor halves wholes strips or minisNo matter what size your Western blot is theres a box that fits it perfectly. With boxes that are correctly matched to common blot sizes you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally all corners are formed at 90 angles and the bottom surface is uniformly flat further reducing consumption of costly reagents and antibodies (for example popular mini-gels require only about 4mL).At a price that makes them truly disposable all sizes are engineered from crystal clear polystyrene. All sizes are also equipped with an easy-to-open hinged lid that reduces evaporation.
Catalog No. 50-290-0543 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7420955EA
Perfect box for every blotReduce antibody usageHinged Lid minimizes evaporationfor halves wholes strips or minisNo matter what size your Western blot is theres a box that fits it perfectly. With boxes that are correctly matched to common blot sizes you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally all corners are formed at 90 angles and the bottom surface is uniformly flat further reducing consumption of costly reagents and antibodies (for example popular mini-gels require only about 4mL).At a price that makes them truly disposable all sizes are engineered from crystal clear polystyrene. All sizes are also equipped with an easy-to-open hinged lid that reduces evaporation.