Learn More
LICORbio Black Western Blot Incubation Boxes (extra large - 5 boxes)
Catalog No. NC9331102
Encompass_Preferred
Black blot boxes are suitable for incubating near-infrared fluorescent and chemiluminescent Western blots or protein gels. The X-Large size is: 15.2 cm x 10.1 cm x 3.1 cm.
Catalog No. NC9331102 Supplier LICORbio Supplier No. 92997405
Black blot boxes are suitable for incubating near-infrared fluorescent and chemiluminescent Western blots or protein gels. The X-Large size is: 15.2 cm x 10.1 cm x 3.1 cm.