Learn More
Electron Microscopy Sciences Western Blotting Boxes XXL, Clear, 12 ml, 6 x 4 x 2"
SDP
Catalog No. 501929219
Encompass_Preferred
No matter what size your Western blot is, there's a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at a 90 deg angle and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Equipped with an easy-to-open hinged lid trhat reduces evaporation. Applications: Bio-Rad Criterion, Amersham.
Color: Clear
Max volume: 12 ml
Dimensions: 6 x 4 x 2"
Catalog No. 50-192-9219 Supplier Electron Microscopy Sciences Supplier No. 7047512
No matter what size your Western blot is, there's a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at a 90 deg angle and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Equipped with an easy-to-open hinged lid trhat reduces evaporation. Applications: Bio-Rad Criterion, Amersham.
Color: Clear
Max volume: 12 ml
Dimensions: 6 x 4 x 2"