Learn More
Electron Microscopy Sciences Blu-Lite UHC™ Western Blotting Film, 8 x 10"
SDP
Catalog No. 501931060
Encompass_Preferred
Blu-Lite UHC high contrast, high sensitivity autoradiography film is specially formulated for an optimized balance of high sensitivity and low background. This low background, high contrast formula produces a lighter blue film, making it ideal for all autoradiographic and electrophoretic applications.
Blu-Lite UHC's is also coated on both sides (double emulsion) for improved consistency and overall performance. Especially useful with chemiluminescence, BluBlot™ HS works well with 14C, 32P, 125I, and 35S. This new film reliably produces sharp images which allow for maximum DNA visualization with unsurpassed clarity.
Compatible with automatic processors and manual development methods.
Size: 8 x 10"
Catalog No. 50-193-1060 Supplier Electron Microscopy Sciences Supplier No. 7047302
Blu-Lite UHC high contrast, high sensitivity autoradiography film is specially formulated for an optimized balance of high sensitivity and low background. This low background, high contrast formula produces a lighter blue film, making it ideal for all autoradiographic and electrophoretic applications.
Blu-Lite UHC's is also coated on both sides (double emulsion) for improved consistency and overall performance. Especially useful with chemiluminescence, BluBlot™ HS works well with 14C, 32P, 125I, and 35S. This new film reliably produces sharp images which allow for maximum DNA visualization with unsurpassed clarity.
Compatible with automatic processors and manual development methods.
Size: 8 x 10"