Learn More
LICORbio Western Blotting Kit I RD (PBS/PVDF)
Catalog No. NC2225455
Encompass_Preferred
This kit contains IRDye 800CW Goat anti-Mouse Secondary Antibody (0.1 mg, 1 mg/mL), IRDye 680RD Goat anti-Rabbit Secondary Antibody (0.1 mg, 1 mg/mL), 500 mL Intercept (PBS) Blocking Buffer, and 10 Immobilon-FL PVDF Membranes (0.45 µm, 10 cm x 10 cm)
This item is not returnable. View return policy
Catalog No. NC2225455 Supplier LICORbio Supplier No. 92632081
This kit contains IRDye 800CW Goat anti-Mouse Secondary Antibody (0.1 mg, 1 mg/mL), IRDye 680RD Goat anti-Rabbit Secondary Antibody (0.1 mg, 1 mg/mL), 500 mL Intercept (PBS) Blocking Buffer, and 10 Immobilon-FL PVDF Membranes (0.45 µm, 10 cm x 10 cm)