Learn More
LICOR Western Blotting Kit IV RD (PBS/NITRO)
Catalog No. NC2225459
Encompass_Preferred
This kit contains IRDye 800CW Goat anti-Rabbit Secondary Antibody (0.1 mg, 1 mg/mL), IRDye 680RD Goat anti-Mouse Secondary Antibody (0.1 mg, 1 mg/mL), 500 mL Intercept (PBS) Blocking Buffer, and 10 Odyssey Nitrocellulose Membranes (0.22 µm, 7 cm x 8.5 cm)
This item is not returnable. View return policy
Catalog No. NC2225459 Supplier LICOR Supplier No. 92632084
This kit contains IRDye 800CW Goat anti-Rabbit Secondary Antibody (0.1 mg, 1 mg/mL), IRDye 680RD Goat anti-Mouse Secondary Antibody (0.1 mg, 1 mg/mL), 500 mL Intercept (PBS) Blocking Buffer, and 10 Odyssey Nitrocellulose Membranes (0.22 µm, 7 cm x 8.5 cm)