Learn More
Advansta Inc WesternBright Sirius Chemiluminescent Detection Kit 200ml
Catalog No. NC0896372
Encompass
WesternBright Sirius is the most sensitive HRP substrate available from Advansta for chemiluminescent Western blotting. With attomole sensitivity and a long-lasting signal, WesternBright Sirius allows you to detect bands not visualized with other substrates. High signal-to-noise and a large dynamic range make it ideal for quantifying low-intensity bands.
Catalog No. NC0896372 Supplier Advansta Inc Supplier No. K12043D20
WesternBright Sirius is the most sensitive HRP substrate available from Advansta for chemiluminescent Western blotting. With attomole sensitivity and a long-lasting signal, WesternBright Sirius allows you to detect bands not visualized with other substrates. High signal-to-noise and a large dynamic range make it ideal for quantifying low-intensity bands.