Learn More
Rockland Immunochemicals WESTERN INCUBATION BOX, LARGE (4 5/8'' X 3 1/2'' X 1 1/8'') , 5-PACK - WIB-4625-005
SDP
Catalog No. NC9893654
Encompass_Preferred
Black plastic hinged box ideal for the incubation, blocking, and washing steps for Western Blotting using Fluorescent Conjugated Antibodies such as DyLight Conjugated Secondary Antibodies or IRDye Antibody Conjugates. This large box is best for volumes between 10mL and 30mL.
Catalog No. NC9893654 Supplier Rockland Immunochemicals Supplier No. WIB4625005
Black plastic hinged box ideal for the incubation, blocking, and washing steps for Western Blotting using Fluorescent Conjugated Antibodies such as DyLight Conjugated Secondary Antibodies or IRDye Antibody Conjugates. This large box is best for volumes between 10mL and 30mL.