Learn More
Genscript Corporation Western Membrane Forceps
Catalog No. 50274533
Encompass
The Western Membrane Forceps are crafted from No. 304 stainless steel. They are resistant to acid, alkali and organic impregnant. You will find these forceps a handy and reliable addition to any study, from small experiments to large projects.
Catalog No. 50-274-533 Supplier Genscript Corporation Supplier No. M00099
The Western Membrane Forceps are crafted from No. 304 stainless steel. They are resistant to acid, alkali and organic impregnant. You will find these forceps a handy and reliable addition to any study, from small experiments to large projects.