Learn More
Enterprise Technology Solutions Western Plastics Pallet Wrap,12 Micron,18"x1500',4/CT,CL
SDP
Catalog No. 5000825631
Encompass_Preferred
Pallet-Tite Pallet Wrap is a multi-layered cast film with a metallocene additive and a high resistance to stretch for superior load retention. 12-micron film (47-gauge) performs at the same level as 80-gauge film to save time and the amount of wrap used for each pallet. Wrap keeps your pallet secure and fully wrapped for optimal security.
Catalog No. 50-008-25631 Supplier Enterprise Technology Solutions Supplier No. WPLHYB58
Pallet-Tite Pallet Wrap is a multi-layered cast film with a metallocene additive and a high resistance to stretch for superior load retention. 12-micron film (47-gauge) performs at the same level as 80-gauge film to save time and the amount of wrap used for each pallet. Wrap keeps your pallet secure and fully wrapped for optimal security.