Learn More
Enterprise Technology Solutions Western Plastics Pallet Wrap,Hand App,Poly,60Gauge,18"x2000',4RL/CT,CL
SDP
Catalog No. 5000823931
Encompass_Preferred
Pallet-Tite Cast Stretch Wrap is designed for hand application to easily wrap pallets. Super-strong, super-cling pallet film is made of 60-gauge linear, low-density polyethylene for thick, secure wrapping. It helps protect against dust, dirt and moisture.
Catalog No. 50-008-23931 Supplier Enterprise Technology Solutions Supplier No. WPLPSF06
Pallet-Tite Cast Stretch Wrap is designed for hand application to easily wrap pallets. Super-strong, super-cling pallet film is made of 60-gauge linear, low-density polyethylene for thick, secure wrapping. It helps protect against dust, dirt and moisture.