Learn More
Enterprise Technology Solutions Western Plastics Stretch Film,Mini,80 Gauge,3"x1000',18/CT,CL
SDP
Catalog No. 5000824227
Encompass_Preferred
EZ Bander Stretch Film is made of ultra-cling material to keep your packages and pallets tightly wrapped. Mini, residue-free film is made of 80-gauge, linear, low-density polyethylene for the ultimate stretch. Easily wrap mini film around packages for simple application.
Catalog No. 50-008-24227 Supplier Enterprise Technology Solutions Supplier No. WPLSBF08CT
EZ Bander Stretch Film is made of ultra-cling material to keep your packages and pallets tightly wrapped. Mini, residue-free film is made of 80-gauge, linear, low-density polyethylene for the ultimate stretch. Easily wrap mini film around packages for simple application.