Learn More
Genhunter Corporation PerfectWestern, Extra Small, Clear
Catalog No. NC9407179
Encompass
Container; Extra Small; PerfectWestern;; 5.3L x 4W x 2.2cm H; Clear; Perfectly accommodate Western blots of different size; Made of durable polystyrene plastic; With ultra-smooth flat inner surfaces and hinged lids to reduce evaporation
Catalog No. NC9407179 Supplier Genhunter Corporation Supplier No. B121
Container; Extra Small; PerfectWestern;; 5.3L x 4W x 2.2cm H; Clear; Perfectly accommodate Western blots of different size; Made of durable polystyrene plastic; With ultra-smooth flat inner surfaces and hinged lids to reduce evaporation