Learn More
LICOR WesternSure PREMIUM Chemiluminescent Substrate (20 mL kit)
Catalog No. NC1079127
Encompass_Preferred
WesternSure PREMIUM Chemiluminescent Substrate is a highly sensitive, enhanced substrate for detecting horseradish peroxidase (HRP) on immunoblots. It has sensitivity comparable with SuperSignal West Dura for greater than 60 minutes post-incubation, it has the most intense signal with longest duration, and it is ideal for use with digital imaging systems. The substrate pack contains Luminol Enhancer Solution and Stable Peroxidase Solution.
Catalog No. NC1079127 Supplier LICOR Supplier No. 92695010
WesternSure PREMIUM Chemiluminescent Substrate is a highly sensitive, enhanced substrate for detecting horseradish peroxidase (HRP) on immunoblots. It has sensitivity comparable with SuperSignal West Dura for greater than 60 minutes post-incubation, it has the most intense signal with longest duration, and it is ideal for use with digital imaging systems. The substrate pack contains Luminol Enhancer Solution and Stable Peroxidase Solution.