Learn More
LICOR WesternSure Pen
Catalog No. NC0705465
Encompass_Preferred
The WesternSure Pen is used to annotate visible protein ladders (such as Chameleon Vue) prior to chemiluminescent Western blot detection. The pen is optimized for detection using the C-DiGit Blot Scanner or the Odyssey Fc Imaging System, and is also suitable for use with film or other imaging systems. This unique marker delivers an ink which emits light when incubated with commonly-used chemiluminescent substrates, including WesternSure PREMIUM Chemiluminescent Substrate. The ink is faintly visible for easy identification of marked membranes.
Catalog No. NC0705465 Supplier LICOR Supplier No. 92691000
The WesternSure Pen is used to annotate visible protein ladders (such as Chameleon Vue) prior to chemiluminescent Western blot detection. The pen is optimized for detection using the C-DiGit Blot Scanner or the Odyssey Fc Imaging System, and is also suitable for use with film or other imaging systems. This unique marker delivers an ink which emits light when incubated with commonly-used chemiluminescent substrates, including WesternSure PREMIUM Chemiluminescent Substrate. The ink is faintly visible for easy identification of marked membranes.