Learn More
LICORbio WesternSure PREMIUM Chemiluminescent Substrate (100 mL kit)
Catalog No. 50489552
Encompass_Preferred
WesternSure PREMIUM Chemiluminescent Substrate is a highly sensitive, enhanced substrate for detecting horseradish peroxidase (HRP) on immunoblots. It has sensitivity comparable with SuperSignal West Dura for greater than 60 minutes post-incubation, it has the most intense signal with longest duration, and it is ideal for use with digital imaging systems. The substrate pack contains Luminol Enhancer Solution and Stable Peroxidase Solution.
Catalog No. 50-489-552 Supplier LICORbio Supplier No. 92695000
WesternSure PREMIUM Chemiluminescent Substrate is a highly sensitive, enhanced substrate for detecting horseradish peroxidase (HRP) on immunoblots. It has sensitivity comparable with SuperSignal West Dura for greater than 60 minutes post-incubation, it has the most intense signal with longest duration, and it is ideal for use with digital imaging systems. The substrate pack contains Luminol Enhancer Solution and Stable Peroxidase Solution.