Learn More
GenDEPOT WestPure PVDF Membrane, 0.45um
Supplier:  GenDEPOT LC7032300
Encompass_Preferred
WestPure PVDF membranes have a higher protein binding capacity than WestPure nitrocellulose membrane. The protein binding capacity of WestPure PVDF ranges from 150-300 ug of protein/cm2 and NitroPure Nitrocellulose ranges from 80-125 ug of protein/cm2. Although PVDF has a higher binding capacity, it could result in increased background in some circumstances.
Catalog No. 50-190-4202