Learn More
Bioworld Westvaco WV3 Medium, 50 L
Catalog No. 50254799
Encompass_Preferred
Westvaco WV3 MediumCauses a significant improvement in shoot micropropagation Contains Macronutrients, Micronutrients, and Viatmins as described by Coke (1996)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L or 50L of mediumPlant tissue culture testedWorking Concentration: 4.29 g/LSoluble in water
Catalog No. 50-254-799 Supplier Bioworld Supplier No. 306301194
Westvaco WV3 MediumCauses a significant improvement in shoot micropropagation Contains Macronutrients, Micronutrients, and Viatmins as described by Coke (1996)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L or 50L of mediumPlant tissue culture testedWorking Concentration: 4.29 g/LSoluble in water