Learn More
Bioworld Westvaco WV5 Medium, 50 L
Catalog No. 50254802
Encompass_Preferred
Westvaco WV5 MediumCauses a significant improvement in the initiation of embryogenic cultures from immature seeds Contains Macronutrients, Micronutrients, and Viatmins as describe by Coke (1996)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L or 50L of mediumWorking Concentration: 5.22 g/LSoluble in water
Catalog No. 50-254-802 Supplier Bioworld Supplier No. 306301204
Westvaco WV5 MediumCauses a significant improvement in the initiation of embryogenic cultures from immature seeds Contains Macronutrients, Micronutrients, and Viatmins as describe by Coke (1996)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L or 50L of mediumWorking Concentration: 5.22 g/LSoluble in water