This wet+dry vacuum is designed to effectively capture and contain fine dust particles. It features a patented triple-layer system that effectively captures 99.97% of particles at 0.5 microns. This versatile machine can handle both wet and dry messes without needing to change filters and is built with durable construction.
- Effectively captures and contains fine dust particles
- Triple-layer filtration captures particles down to 0.5 microns
- External manual filter cleaning maintains suction without opening unit
- True wet+dry operation without changing filters
- Water and slurry-resistant filtration
- Zero dust blowback prevents release of harmful debris
- High-capacity 16-gallon barrel holds up to 40 lb. of dust
- Crush-proof, kink-proof hose
- Tough industrial-grade rubber casters
- Ensures a clean and safe environment