Learn More
MSC Nilfisk Advance Squeegee Assembly, Front Mount, 30 Inch, Wet/Dry Vacuum
Catalog No. 502835589
Encompass
This front-mounted squeegee assembly is designed for efficient wet/dry vacuum operation. With a 30-inch width, it provides effective liquid pickup and is compatible with various wet/dry vacuum models.
  • Front mounted for easy attachment
  • 30 inch squeegee width
  • Constructed from durable plastic
  • ESD safe design
  • Compatible with Summit Pro 18, Viper Shovelnose SN18WD, and SweepScrub SS18WD wet/dry vacuums
Catalog No. 50-283-5589 Supplier MSC Supplier No. 44638583
This front-mounted squeegee assembly is designed for efficient wet/dry vacuum operation. With a 30-inch width, it provides effective liquid pickup and is compatible with various wet/dry vacuum models.
  • Front mounted for easy attachment
  • 30 inch squeegee width
  • Constructed from durable plastic
  • ESD safe design
  • Compatible with Summit Pro 18, Viper Shovelnose SN18WD, and SweepScrub SS18WD wet/dry vacuums