Learn More
MSC Delfin Industrial HV109 PRO HEPA 4520P, 20 gal., 1 ea
Catalog No. 502815554
Encompass
This 'pump-out' solution is designed for simultaneously collecting both wet and dry materials in industrial environments. It features a 2 HP motor, providing powerful airflow and suction, and includes an immersible pump capable of collecting and discharging up to 20 gal. of noncombustible fluids up to 100 ft. away.
  • Rotational molded polyethylene tank resists dents, punctures, and maintains air seal
  • Reliable wet shut-off system prevents liquids from damaging the motor
  • Mesh bag inlet screen captures large particles, preventing flow interruptions
  • Immersible pump with float switch for continuous liquid discharge
  • Two separate power buttons for vacuuming and pump operation
  • Easy to transport even when going up and down stairs with a full tank of liquids
Catalog No. 50-281-5554 Supplier MSC Supplier No. 13675723
This 'pump-out' solution is designed for simultaneously collecting both wet and dry materials in industrial environments. It features a 2 HP motor, providing powerful airflow and suction, and includes an immersible pump capable of collecting and discharging up to 20 gal. of noncombustible fluids up to 100 ft. away.
  • Rotational molded polyethylene tank resists dents, punctures, and maintains air seal
  • Reliable wet shut-off system prevents liquids from damaging the motor
  • Mesh bag inlet screen captures large particles, preventing flow interruptions
  • Immersible pump with float switch for continuous liquid discharge
  • Two separate power buttons for vacuuming and pump operation
  • Easy to transport even when going up and down stairs with a full tank of liquids