Learn More
MSC Ridgid Wet/Dry Vacuum, Electric, 14.0 gal, 11.50 Amp, 1 ea
Catalog No. 502866582
Encompass
This Ridgid wet/dry vacuum is an electric-powered unit designed for efficient cleanup in various environments. It features a durable polyethylene tank and a powerful motor.
  • Electric power source
  • Collection capacity of 14.0 gal
  • Durable polyethylene tank
  • Maximum amperage of 11.50 amp
  • Fine dust cartridge filter
  • Vacuum hose fitting size of 2-1/2 in
  • Includes 2-1/2 in x 10 ft premium locking pro hose
  • Comes with multiple attachments: wet nozzle, floor brush, utility nozzle, crevice tool, two extension wands
Catalog No. 50-286-6582 Supplier MSC Supplier No. 35146497
This Ridgid wet/dry vacuum is an electric-powered unit designed for efficient cleanup in various environments. It features a durable polyethylene tank and a powerful motor.
  • Electric power source
  • Collection capacity of 14.0 gal
  • Durable polyethylene tank
  • Maximum amperage of 11.50 amp
  • Fine dust cartridge filter
  • Vacuum hose fitting size of 2-1/2 in
  • Includes 2-1/2 in x 10 ft premium locking pro hose
  • Comes with multiple attachments: wet nozzle, floor brush, utility nozzle, crevice tool, two extension wands