Learn More
MSC Milwaukee Tool Wet/Dry Vacuum, Battery, 2.0 gal, HEPA Filter
Catalog No. 502837428
Encompass
This Milwaukee Tool Wet/Dry Vacuum is a portable, battery-powered solution for efficient cleanup tasks. Featuring a 2.0-gallon collection capacity and advanced HEPA filtration, it is designed for both wet and dry applications in various environments.
  • Battery-powered for cordless operation
  • HEPA filtration for superior dust and debris capture
  • 2.0 gal collection capacity
  • Bagless vacuum collection for convenient disposal
  • Constructed with a durable plastic tank
  • Provides a maximum air flow of 45 CFM
  • Includes M18 RedLithum XC 4.0 Extended Capacity Battery Pack, wide utility nozzle, wireless vinyl hose, crevice tool, and filter assembly
  • Compact design with an 8.00 in overall width and 1.2 in hose diameter
Catalog No. 50-283-7428 Supplier MSC Supplier No. 36760460
This Milwaukee Tool Wet/Dry Vacuum is a portable, battery-powered solution for efficient cleanup tasks. Featuring a 2.0-gallon collection capacity and advanced HEPA filtration, it is designed for both wet and dry applications in various environments.
  • Battery-powered for cordless operation
  • HEPA filtration for superior dust and debris capture
  • 2.0 gal collection capacity
  • Bagless vacuum collection for convenient disposal
  • Constructed with a durable plastic tank
  • Provides a maximum air flow of 45 CFM
  • Includes M18 RedLithum XC 4.0 Extended Capacity Battery Pack, wide utility nozzle, wireless vinyl hose, crevice tool, and filter assembly
  • Compact design with an 8.00 in overall width and 1.2 in hose diameter