Learn More
IBA LifeSciences WET FRED Set for 5-10 ml columns
Catalog No. NC1938880
Encompass
WET FRED enables the application of large cell culture supernatant or lysate volumes to a gravity flow column in a simple way. Based on the siphon principle, it connects the gravity flow column with the bottle containing culture supernatant, lysate, or buffer that are to be applied.
Catalog No. NC1938880 Supplier IBA LifeSciences Supplier No. 20910001
WET FRED enables the application of large cell culture supernatant or lysate volumes to a gravity flow column in a simple way. Based on the siphon principle, it connects the gravity flow column with the bottle containing culture supernatant, lysate, or buffer that are to be applied.