Learn More
IBA LifeSciences WET FRED Set for 1 ml columns
Catalog No. 502100127
Encompass
WET FRED enables the application of large cell culture supernatant or lysate volumes to a gravity flow column in a simple way. Based on the siphon principle, it connects the gravity flow column with the bottle containing culture supernatant, lysate, or buffer that are to be applied.
Catalog No. 50-210-0127 Supplier IBA LifeSciences Supplier No. 20911001
WET FRED enables the application of large cell culture supernatant or lysate volumes to a gravity flow column in a simple way. Based on the siphon principle, it connects the gravity flow column with the bottle containing culture supernatant, lysate, or buffer that are to be applied.