Learn More
Electron Microscopy Sciences Wet Grip Labels Laser Sheets for Microscope Slides, 22 mm x 22 mm, -80 to 100°C Temperature
SDP
Catalog No. 5033516
Encompass_Preferred
Labels for moist, frosty, or frozen tubes. Features "GRIPS" to wet, frost-covered, and frozen surfaces. Temperature resistant to -80 degrees C. Compatible with centrifuge rotors and resistant to chemicals and solvents. Adheres to plastics, glass, metals, and even paperboard boxes.
Use: Microscope Slides
Size: 22 mm x 22 mm
Temperature: -80 to 100°C
Page: 96 (8 across 12 down)
Catalog No. 50-335-16 Supplier Electron Microscopy Sciences Supplier No. 7759540
Labels for moist, frosty, or frozen tubes. Features "GRIPS" to wet, frost-covered, and frozen surfaces. Temperature resistant to -80 degrees C. Compatible with centrifuge rotors and resistant to chemicals and solvents. Adheres to plastics, glass, metals, and even paperboard boxes.
Use: Microscope Slides
Size: 22 mm x 22 mm
Temperature: -80 to 100°C
Page: 96 (8 across 12 down)