Learn More
Electron Microscopy Sciences Wet Grip Labels On a Roll for 1.5 ml - 2.0 ml tubes, 33 mm x 13 mm, -80 to 100°C Temperature
SDP
Catalog No. 5033518
Encompass_Preferred
Labels for moist, frosty, or frozen tubes. Features "GRIPS" to wet, frost-covered, and frozen surfaces. Temperature resistant to -80 degrees C. Compatible with centrifuge rotors and resistant to chemicals and solvents. Adheres to plastics, glass, metals, and even paperboard boxes.
Use: 1.5 ml - 2.0 ml tubes
Size: 33 mm x 13 mm
Temperature: -80 to 100°C
Catalog No. 50-335-18 Supplier Electron Microscopy Sciences Supplier No. 7759565
Add to Cart
Add to Cart
Labels for moist, frosty, or frozen tubes. Features "GRIPS" to wet, frost-covered, and frozen surfaces. Temperature resistant to -80 degrees C. Compatible with centrifuge rotors and resistant to chemicals and solvents. Adheres to plastics, glass, metals, and even paperboard boxes.
Use: 1.5 ml - 2.0 ml tubes
Size: 33 mm x 13 mm
Temperature: -80 to 100°C