Learn More
Electron Microscopy Sciences Wet Grip Labels Direct Thermal for 1.5 ml tubes, 9.5 mm diameter, -80 to 70°C Temperature
SDP
Catalog No. 5033512
Encompass_Preferred
Labels for moist, frosty, or frozen tubes. Features "GRIPS" to wet, frost-covered, and frozen surfaces. Temperature resistant to -80 degrees C. Compatible with centrifuge rotors and resistant to chemicals and solvents. Adheres to plastics, glass, metals, and even paperboard boxes.
Use: 1.5 ml tubes
Size: 9.5 mm diameter
Temperature: -80 to 70°C
Catalog No. 50-335-12 Supplier Electron Microscopy Sciences Supplier No. 7759460
Labels for moist, frosty, or frozen tubes. Features "GRIPS" to wet, frost-covered, and frozen surfaces. Temperature resistant to -80 degrees C. Compatible with centrifuge rotors and resistant to chemicals and solvents. Adheres to plastics, glass, metals, and even paperboard boxes.
Use: 1.5 ml tubes
Size: 9.5 mm diameter
Temperature: -80 to 70°C