Learn More
OTT Hydromet Adcon - WET - Leaf Wetness Sensor
Catalog No. NC1919664
Encompass_Preferred
The ADCON WET Leaf Wetness Sensor was developed to provide users with a robust and low-maintenance alternative to the traditional style sensors using sheets of paper or human or animal hair. A grid of conductive tracks, etched onto a Teflon-coated ceramic plate reliably simulates the surface of a leaf and outputs a range of readings between 0 and 10. In Adcon’s addVANTAGE Pro software the interpretation of the sensor’s output is based on empirical observation of the behavior of moisture on large-leaved plants. In all disease models currently implemented in addVANTAGE Pro 5.x and 6.x sensor values between 0 and 3 are interpreted as dry, while values from 4 to 10 represent wet. For leaves with particular wetting patterns, or for disease models requiring intermediate steps between wet and dry different sensor drivers can easily be implemented.
Catalog No. NC1919664 Supplier OTT Hydromet Supplier No. 200.733.060
The ADCON WET Leaf Wetness Sensor was developed to provide users with a robust and low-maintenance alternative to the traditional style sensors using sheets of paper or human or animal hair. A grid of conductive tracks, etched onto a Teflon-coated ceramic plate reliably simulates the surface of a leaf and outputs a range of readings between 0 and 10. In Adcon’s addVANTAGE Pro software the interpretation of the sensor’s output is based on empirical observation of the behavior of moisture on large-leaved plants. In all disease models currently implemented in addVANTAGE Pro 5.x and 6.x sensor values between 0 and 3 are interpreted as dry, while values from 4 to 10 represent wet. For leaves with particular wetting patterns, or for disease models requiring intermediate steps between wet and dry different sensor drivers can easily be implemented.