Learn More
Grainger Wet Mop Natural Cotton
Catalog No. 502766344
Encompass_Preferred
The Tough Guy Wet Mop String Mop is a 24 oz. cotton mop head that attaches to a mop handle (sold separately). It has a 4-inch band and features eight plies of cotton yarn for absorbency. This mop head is machine washable for reuse. It can be used for general purpose cleaning of hard floors. The Tough Guy mop head measures 1-1/4" x 4" x 24" and weighs 24 oz. It is made in the USA by Tough Guy of Wisconsin. This mop head offers a economical option for wet mopping floors.
Catalog No. 50-276-6344 Supplier Grainger Supplier No. 16W210
The Tough Guy Wet Mop String Mop is a 24 oz. cotton mop head that attaches to a mop handle (sold separately). It has a 4-inch band and features eight plies of cotton yarn for absorbency. This mop head is machine washable for reuse. It can be used for general purpose cleaning of hard floors. The Tough Guy mop head measures 1-1/4" x 4" x 24" and weighs 24 oz. It is made in the USA by Tough Guy of Wisconsin. This mop head offers a economical option for wet mopping floors.