Learn More
Mopec Wet Powder
SDP
Catalog No. 22039017 Shop All Mopec Products
Change view
Click to view available options
Color:
Black
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-039-017 White
22-039-016 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-039-017 Supplier Mopec Supplier No. FS751
Add to Cart
Add to Cart
Specialized forensic product designed for developing latent fingerprints on adhesive and non-porous surfaces.
  • Performance
    • Develop fingerprints on adhesive and non-porous surfaces
    • Can be used alone or after cyanoacrylate fuming
    • Maintains integrity of delicate or sensitive evidence
    • Performs effectively on moisture-covered surfaces, producing clear, high-contrast fingerprints in challenging conditions.
  • Capacity
    • 250 mL
  • Option and Accessories
    • Available in black (FS750) or white (FS751)
Specifications
Product Type Wet Powder
Color White
Quantity 250 mL