Learn More
Fritsch Milling And Sizing Inc Wet Sieving Kit Analysette 18
SDP
Catalog No. 501534041
Encompass
Consisting of clamping lid with wide spreading spray diffuser, sieve pan with outlet made of stainless steel 400 mm dia. and PVC-hose for wet sieving with ANALYSETTE 18.
Catalog No. 50-153-4041 Supplier Fritsch Milling And Sizing Inc Supplier No. 18.3048.00
Consisting of clamping lid with wide spreading spray diffuser, sieve pan with outlet made of stainless steel 400 mm dia. and PVC-hose for wet sieving with ANALYSETTE 18.