Learn More
Research Products International Corp WetGrip Laser Printer Labels, Microscope Slides
SDP
Catalog No. 501977989
Encompass
Ideal for ice baths, WetGrip Micro-Tube Labels grip to wet or frozen tubes and microscope slides. Labels adhere to glass, plastic, metal and paper. Recommended temperature range is -80°C to 100°C. Supplied on laser printable sheets for printing many labels at once.
Catalog No. 50-197-7989 Supplier Research Products International Corp Supplier No. 247178
Ideal for ice baths, WetGrip Micro-Tube Labels grip to wet or frozen tubes and microscope slides. Labels adhere to glass, plastic, metal and paper. Recommended temperature range is -80°C to 100°C. Supplied on laser printable sheets for printing many labels at once.