Learn More
USA Scientific Inc WetGrip Laser Print Label 0.94 inch x 0.5 inch
Catalog No. NC9846830
Encompass
WetGrip labels on laser printable sheets, white, 0.94 inch x 0.5 in, for 0.5 mL tubes, 2380/box
Catalog No. NC9846830 Supplier USA Scientific Inc Supplier No. 91452380
WetGrip labels on laser printable sheets, white, 0.94 inch x 0.5 in, for 0.5 mL tubes, 2380/box