Learn More
Wexford Labs Inc WEX-CIDE READY TO USE/12QTS/CS
SDP
Supplier:  Wexford Labs Inc 212002
Encompass_Preferred
A Powerful One-Step Cleaner, Disinfectant, And Deodorizer. 12 Per Case. - Use For Disinfection Of All Hard, Non-Porous Surfaces In Healthcare Facilities Including Operating Rooms, Intensive Care, Nurseries, Emergency Areas, And Dental Operatories. - Recommended For Use By Educational, Athletic, And Other Institutional Facilities As Well As Police And Ems To Decontaminate Hard Inanimate Surfaces That Have The Potential To Be Contaminated By Blood Or Other Potentially Infectious Material (Opim) - Easy To Use Non-Aerosol Dispensing Cap On Bottle - Totally Broad Spectrum: Tuberculocidalgermicidal, Pseudomonacidal, Fungicidal And Virucidal - Effective In Blood Serum: Products That Are Effective In The Presence Of Organic Soil Such As 5% Blood Serum Are Classified As One Step Cleaner/Disinfectants (Epa Dis Tss-2) -Effective Against Antibiotic Resistant Organisms, Including Mrsa,Mrse And Vre - Excellent Cleaner / Detergent
Catalog No. NC0660212