Learn More
Medchemexpress LLC HY-133551 5mg Medchemexpress, WF-47-JS03 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1335515MG
Encompass_Preferred
Medchemexpress, HY-133551 5mg WF-47-JS03 CAS: WF-47-JS03 is a potent and selective RET kinase inhibitor with IC50s of 1.7 nM and 5.3 nM for KIF5B-RET transfected Ba/F3 cells and CCDC6-RET transfected LC-2/ad lung cancer cells, respectively. WF-47-JS03 demonstrates >500-fold selectivity against kinase insert domain receptor (KDR). Effective brain penetration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1344