Learn More
Aviva Systems Biology Corp WFDC1 Antibody - middle region (ARP57526_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57526_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WFDC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human WFDC1. Alternate names: PS20ApplicationWB. Species reactivity: Dog, Human, Pig, Rat
Catalog No. 50-014-62851